Product Details

SNP ID
rs146718778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:143121365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTCGTCGTTCTTCGGCGAGCT[A/G]TTCAACCAGAATCCCGAGGTGTTCT
Phenotype
MIM: 603798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST2 PubMed Links

Gene Details

Gene
CHST2
Gene Name
carbohydrate sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004267.4 1488 Silent Mutation CTA,CTG L183L NP_004258.2

View Full Product Details