Product Details

SNP ID
rs147043663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37994284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCCCCCAGAGCCCGAAGGCCAC[A/G]CAGGGGGCGTCCAGCGACCTGCACT
Phenotype
MIM: 608592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
There are no transcripts associated with this gene.

Gene
VILL
Gene Name
villin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015873.3 509 Silent Mutation ACA,ACG T53T NP_056957.3
XM_005265191.3 509 Silent Mutation ACA,ACG T53T XP_005265248.1
XM_005265192.4 509 Silent Mutation ACA,ACG T53T XP_005265249.1
XM_006713184.3 509 Silent Mutation ACA,ACG T53T XP_006713247.1
XM_011533770.2 509 Silent Mutation ACA,ACG T53T XP_011532072.1
XM_011533771.1 509 Silent Mutation ACA,ACG T53T XP_011532073.1
XM_011533773.2 509 Silent Mutation ACA,ACG T53T XP_011532075.1

View Full Product Details