Product Details

SNP ID
rs147072216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:67003703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATGCATACGTTTTCCTCTGATG[A/G]AAGATACCTTTATAGAGAAAATTCC
Phenotype
MIM: 616607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KBTBD8 PubMed Links

Gene Details

Gene
KBTBD8
Gene Name
kelch repeat and BTB domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032505.2 574 Missense Mutation AAA,GAA K246E NP_115894.2
XM_005264771.3 574 Missense Mutation AAA,GAA K169E XP_005264828.1

View Full Product Details