Product Details
- SNP ID
-
rs147270008
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:57757804 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCCACTTTTGATTGCAAAGTGCT[A/G]TCAAGGAACCACGCTCTCGTCTGGT
- Phenotype
-
MIM: 602701
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLMAP
PubMed Links
Gene Details
- Gene
- SLMAP
- Gene Name
- sarcolemma associated protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304420.2 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
NP_001291349.1 |
NM_001304421.2 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
NP_001291350.1 |
NM_001304422.2 |
1379 |
Intron |
|
|
NP_001291351.1 |
NM_001304423.2 |
1379 |
Intron |
|
|
NP_001291352.1 |
NM_001311178.1 |
1379 |
Intron |
|
|
NP_001298107.1 |
NM_001311179.1 |
1379 |
Intron |
|
|
NP_001298108.1 |
NM_007159.4 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
NP_009090.2 |
XM_005265456.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265513.1 |
XM_005265457.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265514.1 |
XM_005265458.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265515.1 |
XM_005265459.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265516.1 |
XM_005265460.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265517.1 |
XM_005265461.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265518.1 |
XM_005265462.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265519.1 |
XM_005265464.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265521.1 |
XM_005265465.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265522.1 |
XM_005265466.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265523.1 |
XM_005265467.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265524.1 |
XM_005265468.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265525.1 |
XM_005265470.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265527.1 |
XM_005265471.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265528.1 |
XM_005265472.3 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_005265529.1 |
XM_006713326.2 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_006713389.1 |
XM_011534103.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_011532405.1 |
XM_011534104.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_011532406.1 |
XM_011534105.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_011532407.1 |
XM_011534106.1 |
1379 |
Intron |
|
|
XP_011532408.1 |
XM_017007146.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862635.1 |
XM_017007147.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862636.1 |
XM_017007148.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862637.1 |
XM_017007149.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862638.1 |
XM_017007150.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862639.1 |
XM_017007151.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862640.1 |
XM_017007152.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862641.1 |
XM_017007153.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862642.1 |
XM_017007154.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862643.1 |
XM_017007155.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862644.1 |
XM_017007156.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862645.1 |
XM_017007157.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862646.1 |
XM_017007158.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862647.1 |
XM_017007159.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862648.1 |
XM_017007160.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862649.1 |
XM_017007161.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862650.1 |
XM_017007162.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862651.1 |
XM_017007163.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862652.1 |
XM_017007164.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862653.1 |
XM_017007165.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862654.1 |
XM_017007166.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862655.1 |
XM_017007167.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862656.1 |
XM_017007168.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862657.1 |
XM_017007169.1 |
1379 |
Silent Mutation |
CTA,CTG |
L51L |
XP_016862658.1 |
XM_017007170.1 |
1379 |
Intron |
|
|
XP_016862659.1 |
View Full Product Details