Product Details

SNP ID
rs147642525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129547511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGCAGGCTGGGGAGCAGCGCC[G/T]GGGCACGTCGGTGGCAGCTATCAAG
Phenotype
MIM: 604282
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
H1FOO PubMed Links

Gene Details

Gene
H1FOO
Gene Name
H1 histone family member O, oocyte specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308262.1 Intron NP_001295191.1
NM_153833.2 Intron NP_722575.1
XM_017005733.1 Intron XP_016861222.1
Gene
PLXND1
Gene Name
plexin D1
There are no transcripts associated with this gene.

View Full Product Details