Product Details

SNP ID
rs147660344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38697592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCAGCTCTGGGCACACATGG[C/G]GTGTTAGAGAGTGCCATGGAGCGGT
Phenotype
MIM: 604427
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCN10A PubMed Links

Gene Details

Gene
SCN10A
Gene Name
sodium voltage-gated channel alpha subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293306.2 5334 Silent Mutation ACC,ACG T1875T NP_001280235.2
NM_001293307.2 5334 Silent Mutation ACC,ACG T1778T NP_001280236.2
NM_006514.3 5334 Silent Mutation ACC,ACG T1876T NP_006505.3
XM_005265371.3 5334 Silent Mutation ACC,ACG T1879T XP_005265428.1
XM_011533993.2 5334 Silent Mutation ACC,ACG T1878T XP_011532295.1
XM_011533994.2 5334 Silent Mutation ACC,ACG T1781T XP_011532296.1

View Full Product Details