Product Details

SNP ID
rs147923543
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51873855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAGTGGCCCTCAATAAAGACAC[A/G]GGAATGGCAGCTGTGCCAGCGCCAA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IQCF5 PubMed Links

Gene Details

Gene
IQCF5
Gene Name
IQ motif containing F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145059.1 378 Missense Mutation CGT,TGT R109C NP_001138531.1
Gene
IQCF5-AS1
Gene Name
IQCF5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details