Product Details

SNP ID
rs148261898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184301538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAGAACTCTTTTCTTTATAGC[A/G]TTTTGCTGCTGACATCATCTCCGTT
Phenotype
MIM: 610145 MIM: 606223
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECE2 PubMed Links

Gene Details

Gene
ECE2
Gene Name
endothelin converting enzyme 2
There are no transcripts associated with this gene.

Gene
PSMD2
Gene Name
proteasome 26S subunit, non-ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278708.1 528 Intron NP_001265637.1
NM_001278709.1 528 Intron NP_001265638.1
NM_002808.4 528 Missense Mutation CAT,CGT H120R NP_002799.3

View Full Product Details