Product Details
- SNP ID
-
rs148424020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:118902559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCCGACTCTGGGCTGGTACCAT[A/G]ACAGGTACTGCACCAATTCGTTCCA
- Phenotype
-
MIM: 608351
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IGSF11
PubMed Links
Gene Details
- Gene
- IGSF11
- Gene Name
- immunoglobulin superfamily member 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001015887.1 |
1504 |
Silent Mutation |
GTC,GTT |
V419V |
NP_001015887.1 |
NM_152538.2 |
1504 |
Silent Mutation |
GTC,GTT |
V418V |
NP_689751.2 |
XM_006713517.1 |
1504 |
Silent Mutation |
GTC,GTT |
V360V |
XP_006713580.1 |
XM_011512462.2 |
1504 |
Silent Mutation |
GTC,GTT |
V469V |
XP_011510764.1 |
XM_011512463.1 |
1504 |
Silent Mutation |
GTC,GTT |
V468V |
XP_011510765.1 |
XM_011512464.2 |
1504 |
Silent Mutation |
GTC,GTT |
V468V |
XP_011510766.1 |
XM_011512465.1 |
1504 |
Silent Mutation |
GTC,GTT |
V468V |
XP_011510767.1 |
XM_011512466.2 |
1504 |
Silent Mutation |
GTC,GTT |
V445V |
XP_011510768.1 |
XM_011512467.2 |
1504 |
Silent Mutation |
GTC,GTT |
V441V |
XP_011510769.1 |
XM_011512468.1 |
1504 |
Silent Mutation |
GTC,GTT |
V431V |
XP_011510770.1 |
XM_011512469.1 |
1504 |
Silent Mutation |
GTC,GTT |
V418V |
XP_011510771.1 |
XM_011512470.1 |
1504 |
Silent Mutation |
GTC,GTT |
V418V |
XP_011510772.1 |
XM_011512471.2 |
1504 |
Silent Mutation |
GTC,GTT |
V417V |
XP_011510773.1 |
XM_011512472.1 |
1504 |
Silent Mutation |
GTC,GTT |
V394V |
XP_011510774.1 |
XM_011512474.2 |
1504 |
Intron |
|
|
XP_011510776.1 |
XM_017005789.1 |
1504 |
Silent Mutation |
GTC,GTT |
V360V |
XP_016861278.1 |
XM_017005790.1 |
1504 |
Silent Mutation |
GTC,GTT |
V336V |
XP_016861279.1 |
View Full Product Details