Product Details

SNP ID
rs148450926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:109178186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTATTGAGCAGCAAGAATGAGAG[C/T]AGAGGGGGAAGCAAAGGTGGCACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00488 PubMed Links

Gene Details

Gene
LINC00488
Gene Name
long intergenic non-protein coding RNA 488
There are no transcripts associated with this gene.

View Full Product Details