Product Details

SNP ID
rs148481971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42548710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCGCTCTGTGCCCAGGGAGTGA[A/G]AGGCAGGTCCAGGGTGGGAAGAAGA
Phenotype
MIM: 604028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC22C PubMed Links

Gene Details

Gene
SEC22C
Gene Name
SEC22 homolog C, vesicle trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201572.1 5792 Intron NP_001188501.1
NM_001201584.1 5792 Intron NP_001188513.1
NM_004206.3 5792 Intron NP_004197.1
NM_032970.3 5792 UTR 3 NP_116752.1
XM_005265533.1 5792 UTR 3 XP_005265590.1
XM_005265534.1 5792 UTR 3 XP_005265591.1
XM_005265535.1 5792 UTR 3 XP_005265592.1
XM_011534212.1 5792 UTR 3 XP_011532514.1
XM_017007438.1 5792 Intron XP_016862927.1
XM_017007439.1 5792 Intron XP_016862928.1

View Full Product Details