Product Details

SNP ID
rs148678130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98283366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCATTTTTAGGTGGCTTCCTCC[A/T]TGCCTTAATTCATGAAGTCCTTATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5H2 PubMed Links

Gene Details

Gene
OR5H2
Gene Name
olfactory receptor family 5 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005482.1 479 Missense Mutation CAT,CTT H160L NP_001005482.1

View Full Product Details