Product Details

SNP ID
rs148882452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179593845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGTCTGAAGAAGCCTTAGGGC[A/T]TCTTCTCTTTCCTGGACAACTTTAT
Phenotype
MIM: 604958 MIM: 611852
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTL6A PubMed Links

Gene Details

Gene
ACTL6A
Gene Name
actin like 6A
There are no transcripts associated with this gene.

Gene
MRPL47
Gene Name
mitochondrial ribosomal protein L47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020409.2 329 Missense Mutation GAA,GAT E151D NP_065142.2
NM_177988.1 329 Missense Mutation GAA,GAT E41D NP_817125.1

View Full Product Details