Product Details

SNP ID
rs149360688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42686147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCGGACTCTCGGCCGACGAG[C/T]TCATCGCCATCATCTCCAGCGACGG
Phenotype
MIM: 608116 MIM: 615340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HHATL PubMed Links

Gene Details

Gene
HHATL
Gene Name
hedgehog acyltransferase-like
There are no transcripts associated with this gene.

Gene
KLHL40
Gene Name
kelch like family member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152393.3 629 Missense Mutation CTC,TTC L177F NP_689606.2

View Full Product Details