Product Details

SNP ID
rs149371008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:68976242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACAGGGAAAAATTCTCTTCAT[A/G]AGGCTAATTTGAAGTTTTTGAAATT
Phenotype
MIM: 614789
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EOGT PubMed Links

Gene Details

Gene
EOGT
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278689.1 3511 UTR 3 NP_001265618.1
NM_173654.2 3511 UTR 3 NP_775925.1
XM_005264743.2 3511 UTR 3 XP_005264800.1
XM_011533599.1 3511 UTR 3 XP_011531901.1
XM_011533600.1 3511 UTR 3 XP_011531902.1
XM_011533602.1 3511 UTR 3 XP_011531904.1
XM_011533603.1 3511 UTR 3 XP_011531905.1
XM_017006204.1 3511 UTR 3 XP_016861693.1
XM_017006205.1 3511 UTR 3 XP_016861694.1
XM_017006206.1 3511 UTR 3 XP_016861695.1
XM_017006207.1 3511 Intron XP_016861696.1
XM_017006208.1 3511 Intron XP_016861697.1
XM_017006209.1 3511 UTR 3 XP_016861698.1

View Full Product Details