Product Details

SNP ID
rs149379959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49859334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCCTACTCAGCTGGCCTCCTCC[C/G]TTTGAGACTCCTGGGCATAACCAGC
Phenotype
MIM: 614993 MIM: 605958
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAMKV PubMed Links

Gene Details

Gene
CAMKV
Gene Name
CaM kinase like vesicle associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320147.1 1876 Missense Mutation ACG,AGG T466R NP_001307076.1
NM_024046.4 1876 Missense Mutation ACG,AGG T497R NP_076951.2
Gene
TRAIP
Gene Name
TRAF interacting protein
There are no transcripts associated with this gene.

View Full Product Details