Product Details

SNP ID
rs149722904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42548143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATATTTATTGTTACAAGAAAAC[C/T]GTAAGAAGCTTGTGTAACATGAACA
Phenotype
MIM: 604028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC22C PubMed Links

Gene Details

Gene
SEC22C
Gene Name
SEC22 homolog C, vesicle trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201572.1 1374 UTR 3 NP_001188501.1
NM_001201584.1 1374 UTR 3 NP_001188513.1
NM_004206.3 1374 UTR 3 NP_004197.1
NM_032970.3 1374 UTR 3 NP_116752.1
XM_005265533.1 1374 UTR 3 XP_005265590.1
XM_005265534.1 1374 UTR 3 XP_005265591.1
XM_005265535.1 1374 UTR 3 XP_005265592.1
XM_011534212.1 1374 UTR 3 XP_011532514.1
XM_017007438.1 1374 UTR 3 XP_016862927.1
XM_017007439.1 1374 UTR 3 XP_016862928.1

View Full Product Details