Product Details

SNP ID
rs149901533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46439459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCTGCCAGCTTCAAATCCTTA[A/G]CCCATGCCTCATTGTTATTTCCTGG
Phenotype
MIM: 150210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTF PubMed Links

Gene Details

Gene
LTF
Gene Name
lactotransferrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199149.1 1930 Missense Mutation GCT,GTT A538V NP_001186078.1
NM_001321121.1 1930 Missense Mutation GCT,GTT A580V NP_001308050.1
NM_001321122.1 1930 Missense Mutation GCT,GTT A569V NP_001308051.1
NM_002343.5 1930 Missense Mutation GCT,GTT A582V NP_002334.2

View Full Product Details