Product Details

SNP ID
rs149967896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:71754934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCGTCACGCTGCTCTTCCTGCT[C/T]CTCTGGGGGCCCTACGTCGTGGCCA
Phenotype
MIM: 609896 MIM: 605187
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4E3 PubMed Links

Gene Details

Gene
EIF4E3
Gene Name
eukaryotic translation initiation factor 4E family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134649.2 885 Intron NP_001128121.1
NM_001134650.1 885 Intron NP_001128122.1
NM_001134651.1 885 Intron NP_001128123.1
NM_001282886.1 885 Intron NP_001269815.1
NM_173359.4 885 Intron NP_775495.1
XM_011533651.2 885 Intron XP_011531953.1
XM_017006278.1 885 Intron XP_016861767.1
XM_017006279.1 885 Intron XP_016861768.1
XM_017006280.1 885 Intron XP_016861769.1
XM_017006281.1 885 Intron XP_016861770.1
XM_017006282.1 885 Intron XP_016861771.1
Gene
GPR27
Gene Name
G protein-coupled receptor 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018971.1 885 Silent Mutation CTC,CTT L295L NP_061844.1

View Full Product Details