Product Details

SNP ID
rs150244244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126989102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGTGCAGGAGGCAGGCAGCATGG[C/T]GGGCGTGCTCATTGCCGGGCCACCG
Phenotype
MIM: 601055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNA1 PubMed Links

Gene Details

Gene
PLXNA1
Gene Name
plexin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032242.3 1204 Missense Mutation GCG,GTG A170V NP_115618.3
XM_011512908.2 1204 Missense Mutation GCG,GTG A188V XP_011511210.1
XM_011512909.1 1204 Missense Mutation GCG,GTG A188V XP_011511211.1

View Full Product Details