Product Details

SNP ID
rs150496722
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:156541183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACCACCTCATCTAAGCTACAC[C/T]GTATTATACCAAGAATGGTGGCAAT
Phenotype
MIM: 601141 MIM: 606213
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KCNAB1 PubMed Links
Additional Information
For this assay, SNP(s) [rs200179482] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KCNAB1
Gene Name
potassium voltage-gated channel subfamily A member regulatory beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308217.1 2728 Intron NP_001295146.1
NM_001308222.1 2728 Intron NP_001295151.1
NM_003471.3 2728 Intron NP_003462.2
NM_172159.3 2728 Intron NP_751891.1
NM_172160.2 2728 Intron NP_751892.1
XM_011513115.2 2728 Intron XP_011511417.1
XM_011513116.2 2728 Intron XP_011511418.1
XM_011513117.2 2728 Intron XP_011511419.1
XM_017007171.1 2728 Intron XP_016862660.1
XM_017007172.1 2728 Intron XP_016862661.1
XM_017007173.1 2728 Intron XP_016862662.1
XM_017007174.1 2728 Intron XP_016862663.1
Gene
SSR3
Gene Name
signal sequence receptor subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308197.1 2728 UTR 3 NP_001295126.1
NM_001308204.1 2728 UTR 3 NP_001295133.1
NM_001308205.1 2728 UTR 3 NP_001295134.1
NM_007107.4 2728 UTR 3 NP_009038.1

View Full Product Details