Product Details

SNP ID
rs150608432
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51955785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGTGGTGCTGCGCACGCCGGGA[C/T]TGCGCGACGCGCTCTACCTGGCGCA
Phenotype
MIM: 606917 MIM: 607726 MIM: 608503
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPR62 PubMed Links

Gene Details

Gene
GPR62
Gene Name
G protein-coupled receptor 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080865.3 472 Silent Mutation CTG,TTG L45L NP_543141.3
Gene
PARP3
Gene Name
poly(ADP-ribose) polymerase family member 3
There are no transcripts associated with this gene.

Gene
PCBP4
Gene Name
poly(rC) binding protein 4
There are no transcripts associated with this gene.

View Full Product Details