Product Details

SNP ID
rs151164953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126988922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCTGCCCCCACGGCCTGGGCA[A/G]TACTGACAACGTCAACAAGCTGCTG
Phenotype
MIM: 601055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLXNA1 PubMed Links

Gene Details

Gene
PLXNA1
Gene Name
plexin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032242.3 1024 Missense Mutation AAT,AGT N110S NP_115618.3
XM_011512908.2 1024 Missense Mutation AAT,AGT N128S XP_011511210.1
XM_011512909.1 1024 Missense Mutation AAT,AGT N128S XP_011511211.1

View Full Product Details