Product Details

SNP ID
rs151275830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170223732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCCAGCTACTTTGGCTTGCCTT[A/G]CTGCTTGTCCAGGTAGAAGTAAATT
Phenotype
MIM: 600539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCI PubMed Links

Gene Details

Gene
PRKCI
Gene Name
protein kinase C iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002740.5 Intron NP_002731.4
XM_017006860.1 Intron XP_016862349.1

View Full Product Details