Product Details
- SNP ID
-
rs138212222
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:20704740 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGGAAGCAAATCCTCATTGTCA[A/G]CCAGTTCTCCAGAGTCTGCAAGAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PACRGL
PubMed Links
Gene Details
- Gene
- PACRGL
- Gene Name
- PARK2 coregulated like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130727.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
NP_001124199.1 |
NM_001258345.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
NP_001245274.1 |
NM_001258346.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
NP_001245275.1 |
NM_001317849.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
NP_001304778.1 |
NM_145048.3 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
NP_659485.1 |
XM_011513780.1 |
354 |
Missense Mutation |
ACC,GCC |
T93A |
XP_011512082.1 |
XM_011513781.1 |
354 |
Missense Mutation |
ACC,GCC |
T93A |
XP_011512083.1 |
XM_011513784.1 |
354 |
Missense Mutation |
ACC,GCC |
T93A |
XP_011512086.1 |
XM_011513785.1 |
354 |
Missense Mutation |
ACC,GCC |
T66A |
XP_011512087.1 |
XM_011513786.2 |
354 |
Missense Mutation |
ACC,GCC |
T93A |
XP_011512088.1 |
XM_011513787.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512089.1 |
XM_011513788.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512090.1 |
XM_011513791.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512093.1 |
XM_011513792.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512094.1 |
XM_011513794.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512096.1 |
XM_011513795.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512097.1 |
XM_011513796.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512098.1 |
XM_011513797.2 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512099.1 |
XM_011513798.2 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512100.1 |
XM_011513804.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512106.1 |
XM_011513805.2 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_011512107.1 |
XM_017007747.1 |
354 |
Missense Mutation |
ACC,GCC |
T93A |
XP_016863236.1 |
XM_017007748.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863237.1 |
XM_017007749.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863238.1 |
XM_017007750.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863239.1 |
XM_017007751.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863240.1 |
XM_017007752.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863241.1 |
XM_017007753.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863242.1 |
XM_017007754.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863243.1 |
XM_017007755.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863244.1 |
XM_017007756.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863245.1 |
XM_017007757.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863246.1 |
XM_017007758.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863247.1 |
XM_017007759.1 |
354 |
Missense Mutation |
ACC,GCC |
T45A |
XP_016863248.1 |
View Full Product Details