Product Details

SNP ID
rs138800982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5526252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAGATAAAACAGAGCCAGAAT[A/G]CTTTGCTGCCAGCTGGACCTTTGAC
Phenotype
MIM: 607261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVC2 PubMed Links

Gene Details

Gene
EVC2
Gene Name
EvC ciliary complex subunit 2
There are no transcripts associated with this gene.

Gene
LINC01587
Gene Name
long intergenic non-protein coding RNA 1587
There are no transcripts associated with this gene.

View Full Product Details