Product Details

SNP ID
rs139158966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86823730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCCAACTGGCTCGAGAGCCCT[C/G]TGGCAATCCATTGGCCCTGGTGGCC
Phenotype
MIM: 600267 MIM: 613366
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTPN13 PubMed Links

Gene Details

Gene
PTPN13
Gene Name
protein tyrosine phosphatase, non-receptor type 13
There are no transcripts associated with this gene.

Gene
SLC10A6
Gene Name
solute carrier family 10 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197965.2 1240 Missense Mutation CAC,CAG H364Q NP_932069.1

View Full Product Details