Product Details

SNP ID
rs139768582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7060970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCTGGCTCCTTCCCCTCAAC[C/T]GGTGTGTGGAACAAGGCCTCATGTT
Phenotype
MIM: 606173 MIM: 608790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRPEL1 PubMed Links

Gene Details

Gene
GRPEL1
Gene Name
GrpE like 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025196.3 724 Silent Mutation CCA,CCG P182P NP_079472.1
Gene
TADA2B
Gene Name
transcriptional adaptor 2B
There are no transcripts associated with this gene.

View Full Product Details