Product Details

SNP ID
rs140751102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:189941015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGAAGTTTCCCGGAGCCATGGC[C/G]GAGTACTCCTACGTGAAGTCTACCA
Phenotype
MIM: 601278
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FRG1 PubMed Links

Gene Details

Gene
FRG1
Gene Name
FSHD region gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004477.2 Intron NP_004468.1
XM_017007958.1 Intron XP_016863447.1
Gene
LOC728339
Gene Name
uncharacterized LOC728339
There are no transcripts associated with this gene.

View Full Product Details