Product Details

SNP ID
rs141250135
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76163295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACCAAAGAACACACCCAGCGCC[A/G]TGATGATGTAGGGTATGTTGGTGAT
Phenotype
MIM: 602257
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SCARB2 PubMed Links

Gene Details

Gene
SCARB2
Gene Name
scavenger receptor class B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204255.1 1255 Missense Mutation ACG,ATG T300M NP_001191184.1
NM_005506.3 1255 Missense Mutation ACG,ATG T443M NP_005497.1

View Full Product Details