Product Details

SNP ID
rs142587773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188002966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGTGGGCCTTATGTGATGGCTA[C/T]GTGTGCTATGAGCCTGGCCCTCAGG
Phenotype
MIM: 614572
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP42 PubMed Links

Gene Details

Gene
ZFP42
Gene Name
ZFP42 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304358.1 401 Silent Mutation TAC,TAT Y53Y NP_001291287.1
NM_174900.4 401 Intron NP_777560.2
XM_011531604.2 401 Silent Mutation TAC,TAT Y53Y XP_011529906.1
XM_011531605.1 401 Silent Mutation TAC,TAT Y53Y XP_011529907.1
XM_011531606.1 401 Silent Mutation TAC,TAT Y53Y XP_011529908.1
XM_011531607.2 401 Silent Mutation TAC,TAT Y53Y XP_011529909.1

View Full Product Details