Product Details

SNP ID
rs142775374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186194567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGCCGCTGCTGAAGCTCTGGGT[C/T]GGGCCAGTGCCCATGGTGGCCCTTT
Phenotype
MIM: 608614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4V2 PubMed Links

Gene Details

Gene
CYP4V2
Gene Name
cytochrome P450 family 4 subfamily V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207352.3 371 Silent Mutation GTC,GTT V94V NP_997235.3
XM_005262935.3 371 Silent Mutation GTC,GTT V94V XP_005262992.1
XM_017008037.1 371 UTR 5 XP_016863526.1
Gene
FLJ38576
Gene Name
uncharacterized LOC651430
There are no transcripts associated with this gene.

View Full Product Details