Product Details

SNP ID
rs144200936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139061963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGAATCCACCAGCTTGAATAT[A/G]CCTTTTTCTCTCTGAGTCCATTTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELF2 PubMed Links

Gene Details

Gene
ELF2
Gene Name
E74 like ETS transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276457.1 752 Silent Mutation NP_001263386.1
NM_001276458.1 752 Silent Mutation NP_001263387.1
NM_001276459.1 752 Silent Mutation NP_001263388.1
NM_006874.3 752 Silent Mutation NP_006865.1
NM_201999.2 752 Silent Mutation NP_973728.1
XM_005262803.2 752 Silent Mutation XP_005262860.1
XM_005262804.2 752 Silent Mutation XP_005262861.1
XM_005262805.2 752 Silent Mutation XP_005262862.1
XM_006714128.2 752 Silent Mutation XP_006714191.1
XM_006714129.3 752 Silent Mutation XP_006714192.1
XM_011531710.2 752 Silent Mutation XP_011530012.1
XM_011531711.2 752 Silent Mutation XP_011530013.1
XM_011531712.2 752 Silent Mutation XP_011530014.1
XM_011531714.2 752 UTR 5 XP_011530016.1
XM_017007856.1 752 Silent Mutation XP_016863345.1
XM_017007857.1 752 Silent Mutation XP_016863346.1
XM_017007858.1 752 Silent Mutation XP_016863347.1
XM_017007859.1 752 Silent Mutation XP_016863348.1
XM_017007860.1 752 Silent Mutation XP_016863349.1
XM_017007861.1 752 Silent Mutation XP_016863350.1

View Full Product Details