Product Details

SNP ID
rs144293712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99901124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCGGAGTCGATCATCACGGTA[C/T]ACTTTTGCTGCATACTGCATATCTG
Phenotype
MIM: 603296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031723.3 1583 Silent Mutation GTA,GTG V348V NP_001026893.1
NM_001278310.1 1583 Silent Mutation GTA,GTG V281V NP_001265239.1
NM_001278311.1 1583 Intron NP_001265240.1
XM_011532262.2 1583 Silent Mutation GTA,GTG V263V XP_011530564.1
XM_011532263.2 1583 Silent Mutation GTA,GTG V263V XP_011530565.1
XM_011532265.2 1583 Silent Mutation GTA,GTG V260V XP_011530567.1
XM_017008628.1 1583 Silent Mutation GTA,GTG V260V XP_016864117.1
Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
There are no transcripts associated with this gene.

View Full Product Details