Product Details

SNP ID
rs144742463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15612333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGACATGCTGAAACCAAATCCTG[C/T]AGGCCTGCACCAGTTATAGTAAGAC
Phenotype
MIM: 605655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL5 PubMed Links

Gene Details

Gene
FBXL5
Gene Name
F-box and leucine rich repeat protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193534.1 1986 Silent Mutation CTA,CTG L643L NP_001180463.1
NM_001193535.1 1986 Silent Mutation CTA,CTG L627L NP_001180464.1
NM_012161.3 1986 Silent Mutation CTA,CTG L644L NP_036293.1
XM_006713959.3 1986 Silent Mutation CTA,CTG L625L XP_006714022.1
XM_011513831.2 1986 Silent Mutation CTA,CTG L608L XP_011512133.1
XM_011513832.2 1986 Silent Mutation CTA,CTG L589L XP_011512134.1
XM_011513833.2 1986 Silent Mutation CTA,CTG L572L XP_011512135.1
XM_017008019.1 1986 Silent Mutation CTA,CTG L589L XP_016863508.1

View Full Product Details