Product Details

SNP ID
rs145736615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83302201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCGAAGCTTCCTTCTCTTTGAA[A/C]CTTGCACGCTTGCCATTAACACCTT
Phenotype
MIM: 604724
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HPSE PubMed Links

Gene Details

Gene
HPSE
Gene Name
heparanase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098540.2 1189 Missense Mutation GGT,GTT G425V NP_001092010.1
NM_001166498.2 1189 Missense Mutation GGT,GTT G351V NP_001159970.1
NM_001199830.1 1189 Missense Mutation GGT,GTT G367V NP_001186759.1
NM_006665.5 1189 Missense Mutation GGT,GTT G425V NP_006656.2

View Full Product Details