Product Details

SNP ID
rs145934164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15687280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCGAATGAAAGCTTAAAACCTTC[A/G]AAATTAAAAAGGCACTTAGAAACTC
Phenotype
MIM: 600387 MIM: 605655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BST1 PubMed Links

Gene Details

Gene
BST1
Gene Name
bone marrow stromal cell antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004334.2 1282 Intron NP_004325.2
XM_005248185.1 1282 Intron XP_005248242.1
XM_005248186.1 1282 Intron XP_005248243.1
XM_011513878.2 1282 Intron XP_011512180.1
XM_011513879.1 1282 Intron XP_011512181.1
XM_011513881.2 1282 Intron XP_011512183.1
XM_017008565.1 1282 Intron XP_016864054.1
XM_017008566.1 1282 Intron XP_016864055.1
XM_017008567.1 1282 Intron XP_016864056.1
Gene
FAM200B
Gene Name
family with sequence similarity 200 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145191.1 1282 Silent Mutation TCA,TCG S101S NP_001138663.1
XM_017008048.1 1282 Silent Mutation TCA,TCG S101S XP_016863537.1
Gene
FBXL5
Gene Name
F-box and leucine rich repeat protein 5
There are no transcripts associated with this gene.

View Full Product Details