Product Details

SNP ID
rs146037638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185162867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTGGCCACTGGAGGGGTCAAC[C/T]GCTGATCGCTCACTCCTACAACTGA
Phenotype
MIM: 616047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP97 PubMed Links

Gene Details

Gene
CFAP97
Gene Name
cilia and flagella associated protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292033.1 1677 Intron NP_001278962.1
NM_020827.2 1677 Silent Mutation GCA,GCG A510A NP_065878.1
XM_017008483.1 1677 Silent Mutation GCA,GCG A510A XP_016863972.1
XM_017008484.1 1677 Silent Mutation GCA,GCG A510A XP_016863973.1
XM_017008485.1 1677 Silent Mutation GCA,GCG A510A XP_016863974.1
XM_017008486.1 1677 Silent Mutation GCA,GCG A510A XP_016863975.1

View Full Product Details