Product Details
- SNP ID
-
rs146370881
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:122454664 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATCTCAGAATTTGACCACATAA[A/G]TAGGTAGAGCCAGAATTAGATTTCA
- Phenotype
-
MIM: 147680
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL2
PubMed Links
Gene Details
- Gene
- IL2
- Gene Name
- interleukin 2
View Full Product Details