Product Details

SNP ID
rs146622998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:81433263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAGTCATTATTGTAAATACT[A/G]TCAATCTACTATGAGAAACTGTTAA
Phenotype
MIM: 614532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASGEF1B PubMed Links

Gene Details

Gene
RASGEF1B
Gene Name
RasGEF domain family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300735.1 Intron NP_001287664.1
NM_001300736.1 Intron NP_001287665.1
NM_152545.2 Intron NP_689758.1
XM_017007812.1 Intron XP_016863301.1
XM_017007813.1 Intron XP_016863302.1

View Full Product Details