Product Details

SNP ID
rs147182750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121818141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGAGACAAGGCTTAAGACTTT[C/G]CAGGGTATATCCAGTCTTTCGTATT
Phenotype
MIM: 607590 MIM: 123835 MIM: 606180
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BBS7 PubMed Links

Gene Details

Gene
BBS7
Gene Name
Bardet-Biedl syndrome 7
There are no transcripts associated with this gene.

Gene
CCNA2
Gene Name
cyclin A2
There are no transcripts associated with this gene.

Gene
EXOSC9
Gene Name
exosome component 9
There are no transcripts associated with this gene.

View Full Product Details