Product Details

SNP ID
rs147719473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:150583929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGTCGTGCCTGGGCGACCGGCT[C/T]AACGGCATCCTGCTGCAGCTCATCT
Phenotype
MIM: 606453 MIM: 604357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC729558 PubMed Links

Gene Details

Gene
LOC729558
Gene Name
uncharacterized LOC729558
There are no transcripts associated with this gene.

Gene
LRBA
Gene Name
LPS responsive beige-like anchor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199282.2 2005 Intron NP_001186211.2
NM_006726.4 2005 Intron NP_006717.2
XM_005263372.3 2005 Intron XP_005263429.1
XM_005263373.2 2005 Intron XP_005263430.1
XM_005263374.3 2005 Intron XP_005263431.1
XM_005263375.3 2005 Intron XP_005263432.1
XM_011532434.2 2005 Intron XP_011530736.1
XM_017008872.1 2005 Intron XP_016864361.1
XM_017008873.1 2005 Intron XP_016864362.1
XM_017008874.1 2005 Intron XP_016864363.1
Gene
MAB21L2
Gene Name
mab-21 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006439.4 2005 Silent Mutation CTC,CTT L300L NP_006430.1

View Full Product Details