Product Details

SNP ID
rs148664329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7045881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACATCTATGTGGTTTTGGAAG[A/G]AACCAGGCTAGAAGTGGAGAGGCCT
Phenotype
MIM: 608790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC96 PubMed Links

Gene Details

Gene
CCDC96
Gene Name
coiled-coil domain containing 96
There are no transcripts associated with this gene.

Gene
LOC100129931
Gene Name
uncharacterized LOC100129931
There are no transcripts associated with this gene.

Gene
TADA2B
Gene Name
transcriptional adaptor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152293.2 Intron NP_689506.2
XM_011513595.1 Intron XP_011511897.1

View Full Product Details