Product Details

SNP ID
rs148818503
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174237481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTACAAAATCTTCACTAATATT[C/T]TGAGCAGCGCGACGGGTATTTCGAA
Phenotype
MIM: 605649
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FBXO8 PubMed Links

Gene Details

Gene
FBXO8
Gene Name
F-box protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012180.2 1741 Silent Mutation CAA,CAG Q297Q NP_036312.2
XM_017008020.1 1741 Silent Mutation CAA,CAG Q150Q XP_016863509.1

View Full Product Details