Product Details

SNP ID
rs148890991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41939340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTTAGAGGACAGCTGCCACTA[C/T]CTGTTGTATTCACTCATCTTTGTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM33 PubMed Links

Gene Details

Gene
TMEM33
Gene Name
transmembrane protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018126.2 437 Silent Mutation TAC,TAT Y95Y NP_060596.2
XM_005248116.3 437 Silent Mutation TAC,TAT Y95Y XP_005248173.1
XM_005248117.2 437 Silent Mutation TAC,TAT Y95Y XP_005248174.1

View Full Product Details