Product Details

SNP ID
rs150462553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75516838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAAAAGACTTGATGTGAATGC[A/G]GCCGGCATTTGGGAGCCTAAAAAAG
Phenotype
MIM: 607680 MIM: 612535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCHY1 PubMed Links

Gene Details

Gene
RCHY1
Gene Name
ring finger and CHY zinc finger domain containing 1
There are no transcripts associated with this gene.

Gene
THAP6
Gene Name
THAP domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317791.1 257 Silent Mutation GCA,GCG A49A NP_001304720.1
NM_144721.5 257 Silent Mutation GCA,GCG A49A NP_653322.1
XM_005262772.2 257 Silent Mutation GCA,GCG A8A XP_005262829.1
XM_005262774.3 257 Silent Mutation GCA,GCG A49A XP_005262831.1
XM_006714109.3 257 Silent Mutation GCA,GCG A49A XP_006714172.1
XM_011531666.2 257 Silent Mutation GCA,GCG A49A XP_011529968.1
XM_011531667.2 257 Silent Mutation GCA,GCG A49A XP_011529969.1
XM_017007800.1 257 Silent Mutation GCA,GCG A49A XP_016863289.1
XM_017007801.1 257 Silent Mutation GCA,GCG A49A XP_016863290.1

View Full Product Details