Product Details

SNP ID
rs150567744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:95840615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGGATCGATGCATATGTATAC[C/G]AAGAACTTCTATGGGGGCAATGGCA
Phenotype
MIM: 179061
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDHA2 PubMed Links

Gene Details

Gene
PDHA2
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005390.4 528 Silent Mutation ACC,ACG T155T NP_005381.1

View Full Product Details