Product Details

SNP ID
rs151091131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:95840396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGCAGATCAGCTGTACAAACA[G/T]AAATTCATTCGCGGTTTCTGTCACC
Phenotype
MIM: 179061
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDHA2 PubMed Links

Gene Details

Gene
PDHA2
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005390.4 309 Silent Mutation CAG,CAT Q82H NP_005381.1

View Full Product Details