Product Details

SNP ID
rs138368479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157463148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAAGTCCTGTGCCAGATTGTCA[A/G]TGACCTCAGCCCTGAGGTGCCCAGC
Phenotype
MIM: 609383
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724404 PubMed Links

Gene Details

Gene
LOC102724404
Gene Name
uncharacterized LOC102724404
There are no transcripts associated with this gene.

Gene
NIPAL4
Gene Name
NIPA like domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099287.1 394 Missense Mutation AAT,AGT N93S NP_001092757.1
NM_001172292.1 394 Missense Mutation AAT,AGT N93S NP_001165763.1
XM_011534552.1 394 UTR 5 XP_011532854.1

View Full Product Details