Product Details

SNP ID
rs138448654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138947265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTGCCGAGCTGGGGGTCCTGA[C/T]GGTAGCGGTCCCGGCAGGTGGTCAG
Phenotype
MIM: 608005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIL1 PubMed Links

Gene Details

Gene
SIL1
Gene Name
SIL1 nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037633.1 1481 Missense Mutation NP_001032722.1
NM_022464.4 1481 Missense Mutation NP_071909.1
XM_011543570.2 1481 Missense Mutation XP_011541872.1

View Full Product Details